Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ditridecyl Phthalate (mixture of branched chain isomers), TCI America™
SDP

Catalog No. P030725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P030725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P030725G Supplier TCI America Supplier No. P030725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 75359-31-8
Molecular Formula C34H58O4
Molecular Weight (g/mol) 530.834
MDL Number MFCD00048451
InChI Key YCZJVRCZIPDYHH-UHFFFAOYSA-N
Synonym Phthalic Acid Ditridecyl Ester
PubChem CID 8379
IUPAC Name ditridecyl benzene-1,2-dicarboxylate
SMILES CCCCCCCCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCCCCCCCC

Specifications

Color Colorless
Quantity 25 g
Formula Weight 530.83
Physical Form Liquid
Chemical Name or Material Ditridecyl Phthalate (mixture of branched chain isomers)
EINECSNumber (3)-1307
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.