Learn More
Cayman Chemical DItyrosInhydrochlorIde 10mg
SDP
Catalog No. 502937863
Encompass_Preferred
A protein oxidation product; intragastric administration decreases hippocampal expression of the NMDA receptor subunits Nr1, Nr2a, and Nr2b and induces memory impairments in a novel object recognition test in mice at 320 UG/kg per day; increases fasting blood glucose levels and decreases plasma insulin levels and pancreatic expression of the insulin synthesis-related genes Ins2, Pdx1, and MafA in mice; increased levels are positively correlated with various diseases
Catalog No. 50-293-7863 Supplier Cayman Chemical Supplier No. 2765710 MG
A protein oxidation product; intragastric administration decreases hippocampal expression of the NMDA receptor subunits Nr1, Nr2a, and Nr2b and induces memory impairments in a novel object recognition test in mice at 320 UG/kg per day; increases fasting blood glucose levels and decreases plasma insulin levels and pancreatic expression of the insulin synthesis-related genes Ins2, Pdx1, and MafA in mice; increased levels are positively correlated with various diseases