Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diundecylamine 97.0+%, TCI America™
SDP

Catalog No. D27695G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D27695G Supplier TCI America Supplier No. D27695G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16165-33-6
Molecular Formula C22H47N
Molecular Weight (g/mol) 325.625
MDL Number MFCD00026548
InChI Key NKGSHSILLGXYDW-UHFFFAOYSA-N
PubChem CID 519223
ChEBI CHEBI:74110
IUPAC Name N-undecylundecan-1-amine
SMILES CCCCCCCCCCCNCCCCCCCCCCC

Specifications

Melting Point 49°C
Color White-Yellow
Boiling Point 232°C
Quantity 5 g
Formula Weight 325.63
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Diundecylamine
EINECSNumber (2)-0138
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.