Learn More
Apexbio Technology LLC Diuron 330-54-1 1g
SDP
Catalog No. 5000089221
Encompass_Preferred
Diuron is a small-molecule inhibitor targeting electron transport within chloroplasts It is designed to interfere with photosynthesis by suppressing electron transfer thereby affecting cellular energy metabolism via reduced ATP and NADH synthesis Diuron exerts its biological activity primarily through inhibition of electron transfer in chloroplasts In experimental studies in animal models dietary administration of Diuron at elevated concentrations is associated with increased occurrence of urothelial carcinoma Based on these pharmacological properties Diuron holds research potential in studies of photosynthetic biochemical pathways cellular energy regulation and chemical-induced carcinogenesis
Catalog No. 50-000-89221 Supplier Apexbio Technology LLC Supplier No. C67311000
Diuron is a small-molecule inhibitor targeting electron transport within chloroplasts It is designed to interfere with photosynthesis by suppressing electron transfer thereby affecting cellular energy metabolism via reduced ATP and NADH synthesis Diuron exerts its biological activity primarily through inhibition of electron transfer in chloroplasts In experimental studies in animal models dietary administration of Diuron at elevated concentrations is associated with increased occurrence of urothelial carcinoma Based on these pharmacological properties Diuron holds research potential in studies of photosynthetic biochemical pathways cellular energy regulation and chemical-induced carcinogenesis