Learn More
Medchemexpress LLC Diuron | 330-54-1 | 99.1% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0860500MG
Encompass_Preferred
Diuron is an orally active phenylurea herbicide that inhibits photosynthesis in plants by blocking the formation of ATP and NADH. It promotes DMBA/BBN-induced bladder cancer and can be utilized in breast cancer research. It increases the production of reactive oxygen species (ROS) and the expression of p53 in certain cell lines. It exhibits herbicidal activity against annual and perennial broadleaf and grass weeds.
  • Significantly increases ROS production in BeWo and MCF-7 cells
  • Induces DNA damage in MCF-7 cells
  • Inhibits *Allium cepa* root tip cell division and causes mitotic abnormalities
  • Can induce urothelial hyperplasia in bladder and renal pelvis in Wistar rats
  • Promotes bladder cancer occurrence in female Swiss mice
Catalog No. 50-004-08551