Learn More
Medchemexpress LLC Diuron-d6 | 1007536-67-5 | 99.5% | 1 MG
SDP
Catalog No. 5000409417
Encompass_Preferred
Diuron-d6 is a deuterium-labeled form of Diuron, an orally active phenylurea herbicide. It functions by inhibiting photosynthesis in plants, specifically by blocking the formation of ATP and NADH. Diuron-d6 also increases the production of Reactive Oxygen Species (ROS) and the expression of p53 in certain cell lines, and has shown herbicidal activity against various broadleaf and grass weeds. It has been noted to promote bladder cancer in some studies and is applicable in breast cancer research.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-09417 Supplier Medchemexpress LLC Supplier No. HYB0860S1MG
Diuron-d6 is a deuterium-labeled form of Diuron, an orally active phenylurea herbicide. It functions by inhibiting photosynthesis in plants, specifically by blocking the formation of ATP and NADH. Diuron-d6 also increases the production of Reactive Oxygen Species (ROS) and the expression of p53 in certain cell lines, and has shown herbicidal activity against various broadleaf and grass weeds. It has been noted to promote bladder cancer in some studies and is applicable in breast cancer research.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS