Learn More
Medchemexpress LLC Diuron-d6 | 1007536-67-5 | 99.5% | 5 MG
SDP
Catalog No. 5000408853
Encompass_Preferred
Diuron-d6 is a deuterium-labeled version of Diuron, an orally active phenylurea herbicide. It functions by inhibiting photosynthesis in plants, specifically by blocking the formation of ATP and NADH. The compound also increases the production of Reactive Oxygen Species (ROS) and has been shown to increase p53 expression in certain cell lines. Diuron-d6 exhibits herbicidal activity against various annual and perennial broadleaf and grass weeds. Additionally, it is noted to promote DMBA/BBN-induced bladder cancer and can be utilized in breast cancer research.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-08853 Supplier Medchemexpress LLC Supplier No. HYB0860S5MG
Add to Cart
Add to Cart
Diuron-d6 is a deuterium-labeled version of Diuron, an orally active phenylurea herbicide. It functions by inhibiting photosynthesis in plants, specifically by blocking the formation of ATP and NADH. The compound also increases the production of Reactive Oxygen Species (ROS) and has been shown to increase p53 expression in certain cell lines. Diuron-d6 exhibits herbicidal activity against various annual and perennial broadleaf and grass weeds. Additionally, it is noted to promote DMBA/BBN-induced bladder cancer and can be utilized in breast cancer research.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.