Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diuron, Spectrum™ Chemical
SDP

Catalog No. 18620682 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-682 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-620-682 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D3925100GM

Synonym: 3-(3,4-Dichlorophenyl)-1,1-dimethylurea, DCMU, Duran

Chemical Identifiers

CAS 330-54-1
Molecular Weight (g/mol) 233.1 g/mol

Specifications

Packaging Bottle
CAS Max % 100
Quantity 100 g
Percent Purity 98%
Chemical Name or Material Diuron
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.