Learn More
Biocare Medical, LLC Diva Decloaker, RTU - 1 gal
SDP
Catalog No. 5082828
Encompass_Preferred
Diva Decloaker is a heat retrieval solution that is compatible with virtually all antibodies and eliminates the need for multiple buffers including citrate buffer, EDTA or high pH Tris buffers. Diva Decloaker can be used with Biocare digital electric pressure cooker (Decloaking Chamber), a steamer, a water bath or a microwave oven. Antibody titers are doubled and tripled when compared to citrate buffer, pH 6.0. Diva Decloaker incorporates Assure technology, a color-coded high temperature pH indicator solution. The end-user is assured by visual inspection that the solution is at the correct dilution and pH. This product is specially formulated for superior pH stability at high temperatures and will help prevent the possibility of losing pH sensitive antigens. Diva Decloaker is non-toxic, non-flammable, odorless and sodium azide and thimerosal free.
Catalog No. 50-828-28 Supplier Biocare Medical, LLC Supplier No. DV2004G1
Diva Decloaker is a heat retrieval solution that is compatible with virtually all antibodies and eliminates the need for multiple buffers including citrate buffer, EDTA or high pH Tris buffers. Diva Decloaker can be used with Biocare digital electric pressure cooker (Decloaking Chamber), a steamer, a water bath or a microwave oven. Antibody titers are doubled and tripled when compared to citrate buffer, pH 6.0. Diva Decloaker incorporates Assure technology, a color-coded high temperature pH indicator solution. The end-user is assured by visual inspection that the solution is at the correct dilution and pH. This product is specially formulated for superior pH stability at high temperatures and will help prevent the possibility of losing pH sensitive antigens. Diva Decloaker is non-toxic, non-flammable, odorless and sodium azide and thimerosal free.