Learn More
Apexbio Technology LLC Divalproex Sodium 76584-70-8 50mg
SDP
Catalog No. 5000091364
Encompass_Preferred
Divalproex Sodium (CAS 76584-70-8) is a small-molecule inhibitor targeting histone deacetylases (HDACs) and modulating GABAergic neurotransmission It is designed to inhibit HDACs and modulate associated signaling pathways thereby regulating neuronal excitability and gene expression Divalproex Sodium exerts its biological activity primarily through HDAC inhibition enhancement of GABA levels inhibition of voltage-gated sodium channels and modulation of NMDA receptor-mediated glutamatergic signaling In experimental studies Divalproex Sodium demonstrates HDAC inhibitory activity with reported IC50 values typically ranging between 400 600 M varying by assay Based on these pharmacological properties Divalproex Sodium holds research potential in epilepsy bipolar disorder and migraine prophylaxis
Catalog No. 50-000-91364 Supplier Apexbio Technology LLC Supplier No. B139150
Divalproex Sodium (CAS 76584-70-8) is a small-molecule inhibitor targeting histone deacetylases (HDACs) and modulating GABAergic neurotransmission It is designed to inhibit HDACs and modulate associated signaling pathways thereby regulating neuronal excitability and gene expression Divalproex Sodium exerts its biological activity primarily through HDAC inhibition enhancement of GABA levels inhibition of voltage-gated sodium channels and modulation of NMDA receptor-mediated glutamatergic signaling In experimental studies Divalproex Sodium demonstrates HDAC inhibitory activity with reported IC50 values typically ranging between 400 600 M varying by assay Based on these pharmacological properties Divalproex Sodium holds research potential in epilepsy bipolar disorder and migraine prophylaxis