Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Divarasib adipate | 2762240-36-6 | 99.2% | C35H42ClF4N7O6 | 1 MG
SDP

Catalog No. 5000355179
Encompass_Preferred

Divarasib (GDC-6036) adipate is an orally bioavailable, highly potent, and selective KRAS G12C inhibitor. It functions by covalently binding to the switch II (SW-II) pocket of KRAS G12C, irreversibly locking it in an inactive GDP-bound state. This product appears as a white to off-white solid.

  • Orally bioavailable
  • Highly potent and selective
  • Functions as a KRAS G12C inhibitor
  • Irreversibly locks KRAS G12C in an inactive state
  • Typical purity of 99.2%
  • Available as a white to off-white solid
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-003-55179 Supplier Medchemexpress LLC Supplier No. HY145928B1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Divarasib (GDC-6036) adipate is an orally bioavailable, highly potent, and selective KRAS G12C inhibitor. It functions by covalently binding to the switch II (SW-II) pocket of KRAS G12C, irreversibly locking it in an inactive GDP-bound state. This product appears as a white to off-white solid.

  • Orally bioavailable
  • Highly potent and selective
  • Functions as a KRAS G12C inhibitor
  • Irreversibly locks KRAS G12C in an inactive state
  • Typical purity of 99.2%
  • Available as a white to off-white solid
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.