Learn More
Medchemexpress LLC Divarasib adipate | 2762240-36-6 | 99.2% | C35H42ClF4N7O6 | 1 ML
SDP
Catalog No. 5000354448
Encompass_Preferred
Divarasib (GDC-6036) adipate is an orally bioavailable, highly potent, and selective KRAS G12C inhibitor. It covalently binds to the switch II (SW-II) pocket of KRAS G12C and irreversibly locks it in the inactive GDP-bound state.
  • Highly potent and selective KRAS G12C inhibitor
  • Orally bioavailable
  • Covalently binds to the switch II (SW-II) pocket of KRAS G12C
  • Irreversibly locks KRAS G12C in the inactive GDP-bound state
Catalog No. 50-003-54448 Supplier Medchemexpress LLC Supplier No. HY145928B10MM/1ML
Divarasib (GDC-6036) adipate is an orally bioavailable, highly potent, and selective KRAS G12C inhibitor. It covalently binds to the switch II (SW-II) pocket of KRAS G12C and irreversibly locks it in the inactive GDP-bound state.
  • Highly potent and selective KRAS G12C inhibitor
  • Orally bioavailable
  • Covalently binds to the switch II (SW-II) pocket of KRAS G12C
  • Irreversibly locks KRAS G12C in the inactive GDP-bound state