Learn More
Medchemexpress LLC Divarasib adipate | 2762240-36-6 | 99.2% | C35H42ClF4N7O6 | 100 MG
SDP
Catalog No. 5000356134
Encompass_Preferred
Divarasib adipate is an orally bioavailable, highly potent, and selective KRAS G12C inhibitor. It covalently binds to the switch II (SW-II) pocket of KRAS G12C, irreversibly locking it in the inactive GDP-bound state.
  • Highly potent and selective KRAS G12C inhibitor
  • Orally bioavailable
  • Locks KRAS G12C in the inactive GDP-bound state
  • White to off-white solid
  • Associated with clinical trials for various conditions
Catalog No. 50-003-56134 Supplier Medchemexpress LLC Supplier No. HY145928B100MG
Divarasib adipate is an orally bioavailable, highly potent, and selective KRAS G12C inhibitor. It covalently binds to the switch II (SW-II) pocket of KRAS G12C, irreversibly locking it in the inactive GDP-bound state.
  • Highly potent and selective KRAS G12C inhibitor
  • Orally bioavailable
  • Locks KRAS G12C in the inactive GDP-bound state
  • White to off-white solid
  • Associated with clinical trials for various conditions