Learn More
Medchemexpress LLC Divarasib adipate | 2762240-36-6 | 99.2% | C35H42ClF4N7O6 | 10MG
SDP
Catalog No. 5000259594
Encompass_Preferred
Divarasib adipate is the adipate salt of divarasib (GDC-6036), a covalent KRAS G12C inhibitor for preclinical research. It is orally bioavailable and highly potent, irreversibly binding the switch II pocket of KRAS G12C to lock the protein in the inactive GDP-bound state. Supplied as a solid with analytical documentation for laboratory use.
  • Highly potent KRAS G12C inhibition (IC50 <0.01 μM).
  • Covalent mechanism locking KRAS G12C in the inactive GDP-bound state.
  • Orally bioavailable small molecule suitable for in vivo and in vitro studies.
  • Provided as the adipate salt for consistent handling and formulation.
  • Delivered with purity data and HPLC documentation for quality assessment.
Catalog No. 50-002-59594 Supplier Medchemexpress LLC Supplier No. HY145928B10MG
Divarasib adipate is the adipate salt of divarasib (GDC-6036), a covalent KRAS G12C inhibitor for preclinical research. It is orally bioavailable and highly potent, irreversibly binding the switch II pocket of KRAS G12C to lock the protein in the inactive GDP-bound state. Supplied as a solid with analytical documentation for laboratory use.
  • Highly potent KRAS G12C inhibition (IC50 <0.01 μM).
  • Covalent mechanism locking KRAS G12C in the inactive GDP-bound state.
  • Orally bioavailable small molecule suitable for in vivo and in vitro studies.
  • Provided as the adipate salt for consistent handling and formulation.
  • Delivered with purity data and HPLC documentation for quality assessment.