Learn More
Medchemexpress LLC Divarasib adipate | 2762240-36-6 | 99.2% | C35H42ClF4N7O6 | 50 MG

Supplier: Medchemexpress LLC HY145928B50MG
Divarasib adipate is an orally bioavailable, highly potent, and selective KRAS G12C inhibitor with an IC50 of <0.01 μM. It covalently binds to the switch II (SW-II) pocket of KRAS G12C, irreversibly locking it in the inactive GDP-bound state. This product is for research use only.
- Highly potent and selective KRAS G12C inhibitor
- Orally bioavailable
- Purity of 99.2%
- Appearance: Solid, white to off-white
- Store powder at -20°C for 3 years or 4°C for 2 years
- Store in solvent at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.