Learn More
Medchemexpress LLC DIVARASIB / GDC-6036 10MG
SDP
Catalog No. NC2584662
Encompass_Preferred
Divarasib (GDC-6036) is an orally bioavailable, highly potent, and selective KRAS G12C inhibitor with an IC50 of <0.01 μM. Divarasib covalently binds to the switch II (SW-II) pocket of KRAS G12C and irreversibly locks it in the inactive GDP-bound state.
Catalog No. NC2584662 Supplier Medchemexpress LLC Supplier No. HY14592810MG
Divarasib (GDC-6036) is an orally bioavailable, highly potent, and selective KRAS G12C inhibitor with an IC50 of <0.01 μM. Divarasib covalently binds to the switch II (SW-II) pocket of KRAS G12C and irreversibly locks it in the inactive GDP-bound state.