Learn More
Enterprise Technology Solutions Diversey Care Aquaria ProX Floor Finish, Ready to Use, 5 Gallon Envirobox, Off-White
SDP
Catalog No. 5000790896
Encompass_Preferred
Aquaria ProX Floor Finish features durable, proven polymer technology that is easy to apply and easy to strip. Clean, long-lasting floor finish meets your environmental expectations while delivering exceptional clarity with outstanding gloss. It also provides an excellent buff response. Ready-to-use floor finish meets Green Seal GS 40 standard and ASTM D-2047 requirement for slip-resistance. It is formulated to work with Pro Strip™ SC Stripper, Attac SC Stripper, Stride Neutral Cleaner and other Diversey Green Seal certified products. Product does not contain any intentionally added PFAS.
Catalog No. 50-007-90896 Supplier Enterprise Technology Solutions Supplier No. DVO101110021
Aquaria ProX Floor Finish features durable, proven polymer technology that is easy to apply and easy to strip. Clean, long-lasting floor finish meets your environmental expectations while delivering exceptional clarity with outstanding gloss. It also provides an excellent buff response. Ready-to-use floor finish meets Green Seal GS 40 standard and ASTM D-2047 requirement for slip-resistance. It is formulated to work with Pro Strip™ SC Stripper, Attac SC Stripper, Stride Neutral Cleaner and other Diversey Green Seal certified products. Product does not contain any intentionally added PFAS.