Learn More
Enterprise Technology Solutions Diversey Care Beer Clean Low Suds Glass Cleaner, .5oz.,
Supplier: Enterprise Technology Solutions DVO990224
Use Beer Clean Low Suds Glass Cleaner for sparkling clean glassware and better-tasting beer. Powdered compound cleans in hot or cold and in hard or soft water. Concentrated formula is designed for quick cleaning with electric brushes. With minimal sudsing, this odorless detergent system provides excellent rinsing results.