Learn More
Enterprise Technology Solutions Diversey Care Beer Clean Mineral Solvent, 14g, 100PK/CT, WE
SDP
Catalog No. 5000790290
Encompass_Preferred
Perfect for behind the counter drinkware cleaning systems, Beer Clean Mineral Solvent serves as a "booster" in the water-softening process and can be especially important in adjusting local water conditions. It ties up hard water minerals to obtain maximum cleaning results. This solvent provides thoroughly cleaned glassware with no odor, aftertaste or residue. Single-use packet allows better dilution control. It is NSF G6 registered for steam table treatment.
Catalog No. 50-007-90290 Supplier Enterprise Technology Solutions Supplier No. DVO990222
Perfect for behind the counter drinkware cleaning systems, Beer Clean Mineral Solvent serves as a "booster" in the water-softening process and can be especially important in adjusting local water conditions. It ties up hard water minerals to obtain maximum cleaning results. This solvent provides thoroughly cleaned glassware with no odor, aftertaste or residue. Single-use packet allows better dilution control. It is NSF G6 registered for steam table treatment.