Learn More
Enterprise Technology Solutions Diversey Care Bio-Floor Plus No-Rinse Floor Cleaner, 1.5L, 2/CT, Green
SDP
Catalog No. 5000790075
Encompass_Preferred
Suma Bio-Floor Plus No-Rinse Floor Cleaner removes fat, grease and oil buildups, cleans soils trapped in tile pores and grout and eliminates odors. Beneficial, self-replicating bacteria continues to clean after mopping is complete. Residual cleaning effect on quarry tile floors can last for up to several days. Surfactant/solvent-based, premium liquid is also safe for use on ceramic tile and vinyl. Apply cost-effective concentrate with mop or spray using innovative RTD spray method.
Catalog No. 50-007-90075 Supplier Enterprise Technology Solutions Supplier No. DVO101102611
Suma Bio-Floor Plus No-Rinse Floor Cleaner removes fat, grease and oil buildups, cleans soils trapped in tile pores and grout and eliminates odors. Beneficial, self-replicating bacteria continues to clean after mopping is complete. Residual cleaning effect on quarry tile floors can last for up to several days. Surfactant/solvent-based, premium liquid is also safe for use on ceramic tile and vinyl. Apply cost-effective concentrate with mop or spray using innovative RTD spray method.