Learn More
Enterprise Technology Solutions Diversey Care Carefree Matte ProX Floor Finish, Low Gloss, 5 Gallon, White
SDP
Catalog No. 5000790531
Encompass_Preferred
Carefree Matte ProX Low Gloss Floor Finish is a versatile, high-performance floor finish without the high gloss. Low-gloss characteristics help hide blemishes, cut lines, traffic lanes and dirt embedment to provide an even, clean look over a longer period of time. Versatile floor finish works with a wide variety of substrates and in many different maintenance methods. It is not intended for use on cork, wood or bamboo flooring. Floor finish meets ASTM D-2047 slip-resistance requirements. Product does not contain any intentionally added PFAS.
Catalog No. 50-007-90531 Supplier Enterprise Technology Solutions Supplier No. DVO101110003
Carefree Matte ProX Low Gloss Floor Finish is a versatile, high-performance floor finish without the high gloss. Low-gloss characteristics help hide blemishes, cut lines, traffic lanes and dirt embedment to provide an even, clean look over a longer period of time. Versatile floor finish works with a wide variety of substrates and in many different maintenance methods. It is not intended for use on cork, wood or bamboo flooring. Floor finish meets ASTM D-2047 slip-resistance requirements. Product does not contain any intentionally added PFAS.