Learn More
Enterprise Technology Solutions Diversey Care Cleaner, Conc, General-Purpose, Citrus, 1 Gal, 2/CT, Green
SDP
Catalog No. 5000790547
Encompass_Preferred
Highly versatile, general-purpose cleaner concentrate contains a blend of high-performance detergents to effectively clean surfaces such as counters, tables and floors. It removes all types of dirt, grit, grease and soap scum with no rinsing required. GP Forward SC General Purpose Cleaner can be used on any water-safe surface and has a pleasant citrus scent. It is excellent for use in scrub and recoat floor-care programs. It works with cold, hot and soft/hard water in mop, bucket, autoscrubber and trigger spray applications. Dilution ratios are 1:64 and 1:128.
Catalog No. 50-007-90547 Supplier Enterprise Technology Solutions Supplier No. DVO101104494
Highly versatile, general-purpose cleaner concentrate contains a blend of high-performance detergents to effectively clean surfaces such as counters, tables and floors. It removes all types of dirt, grit, grease and soap scum with no rinsing required. GP Forward SC General Purpose Cleaner can be used on any water-safe surface and has a pleasant citrus scent. It is excellent for use in scrub and recoat floor-care programs. It works with cold, hot and soft/hard water in mop, bucket, autoscrubber and trigger spray applications. Dilution ratios are 1:64 and 1:128.