Learn More
Enterprise Technology Solutions Diversey Care Cleaner/Degreaser,Conc,RTD,Non-Buty,1.5L/50.7,2/CT,Purple
SDP
Catalog No. 5000790576
Encompass_Preferred
Part of the Diversey RTD System, Speedball Power Cleaner and Degreaser cleans the toughest soils, removing grease, oil, encrusted soil, tire marks, ink, smoke, carbon film, graffiti, heel marks and more. Environmentally preferred, concentrated formula is a non-butyl power cleaner with strong cleaning and degreasing ability. Superior cleaner requires no rinsing and leaves no residue behind. Use as a spray-and-wipe. Dilution ratio is 1:16. NSF certified cleaner/degreaser comes in a special container with an ergonomically designed handle that connects to any convenient water supply via a standard hose connector. Simply select the dilution required, and the cleaner will be accurately and safely dispensed through its Spill-Tite head every time.
Catalog No. 50-007-90576 Supplier Enterprise Technology Solutions Supplier No. DVO95892175
Part of the Diversey RTD System, Speedball Power Cleaner and Degreaser cleans the toughest soils, removing grease, oil, encrusted soil, tire marks, ink, smoke, carbon film, graffiti, heel marks and more. Environmentally preferred, concentrated formula is a non-butyl power cleaner with strong cleaning and degreasing ability. Superior cleaner requires no rinsing and leaves no residue behind. Use as a spray-and-wipe. Dilution ratio is 1:16. NSF certified cleaner/degreaser comes in a special container with an ergonomically designed handle that connects to any convenient water supply via a standard hose connector. Simply select the dilution required, and the cleaner will be accurately and safely dispensed through its Spill-Tite head every time.