Learn More
Enterprise Technology Solutions Diversey Care Cleaner/Degreaser,RTU,Non-Butyl,946ml/32 oz,12/CT,Purple
SDP
Catalog No. 5000790485
Encompass_Preferred
Speedball Power Cleaner and Degreaser is ready to use and cleans the toughest soils, removing grease, oil, encrusted soil, tire marks, ink, smoke, carbon film, graffiti, heel marks and more. Environmentally preferred, concentrated formula is a non-butyl power cleaner with strong cleaning and degreasing ability. Superior cleaner requires no rinsing and leaves no residue behind. Use as a spray-and-wipe.
Catalog No. 50-007-90485 Supplier Enterprise Technology Solutions Supplier No. DVO95891164
Speedball Power Cleaner and Degreaser is ready to use and cleans the toughest soils, removing grease, oil, encrusted soil, tire marks, ink, smoke, carbon film, graffiti, heel marks and more. Environmentally preferred, concentrated formula is a non-butyl power cleaner with strong cleaning and degreasing ability. Superior cleaner requires no rinsing and leaves no residue behind. Use as a spray-and-wipe.