Learn More
Enterprise Technology Solutions Diversey Care Cleaner/Emulsifier,Wiwax,f/Floors,1 Gallon,White
SDP
Catalog No. 5000790533
Encompass_Preferred
Wiwax ProX Maintenance Emulsion is a unique blend of waxes, polymers and detergents to effectively clean and maintain rubber as well as linoleum, asphalt, marble, stone, concrete and slate flooring. It delivers a clean floor with a shiny appearance. This integral part of a comprehensive floor-care program for cleaning, stripping and sealing rubber floors provides protection when used on a regular basis. Varied ratios include 1:1, 1:10, 1:32, 1:64 and 1:128. Arched handle on bottle allows easy pouring. Cleaner does not contain any intentionally added PFAS.
Catalog No. 50-007-90533 Supplier Enterprise Technology Solutions Supplier No. DVO101110064
Wiwax ProX Maintenance Emulsion is a unique blend of waxes, polymers and detergents to effectively clean and maintain rubber as well as linoleum, asphalt, marble, stone, concrete and slate flooring. It delivers a clean floor with a shiny appearance. This integral part of a comprehensive floor-care program for cleaning, stripping and sealing rubber floors provides protection when used on a regular basis. Varied ratios include 1:1, 1:10, 1:32, 1:64 and 1:128. Arched handle on bottle allows easy pouring. Cleaner does not contain any intentionally added PFAS.