Learn More
Enterprise Technology Solutions Diversey Care Cleaner, f/CRT/Plexiglas, 32oz,12/CT,PE
SDP
Catalog No. 5000830521
Encompass_Preferred
Lite Touch CRT and Plexiglas Cleaner dissipates a static charge and repels dust so surfaces remain cleaner longer. It can be used on glass, CRT screens/cabinets, clear plastic windows, Plexiglas, bar-code scanners, hard floors, countertops and many other washable surfaces. Lite Touch is an excellent, solvent-free glass cleaner. It is purple in color with no added fragrance.
Catalog No. 50-008-30521 Supplier Enterprise Technology Solutions Supplier No. DVO03970
Lite Touch CRT and Plexiglas Cleaner dissipates a static charge and repels dust so surfaces remain cleaner longer. It can be used on glass, CRT screens/cabinets, clear plastic windows, Plexiglas, bar-code scanners, hard floors, countertops and many other washable surfaces. Lite Touch is an excellent, solvent-free glass cleaner. It is purple in color with no added fragrance.