Learn More
Enterprise Technology Solutions Diversey Care Cleaner,f/Glass/Multipurpose,Conc,2.5 L J-Fill,Blue
SDP
Catalog No. 5000830675
Encompass_Preferred
Super-concentrated, non-ammoniated glass cleaner meets EcoLogo and Greenguard standards and delivers streak-free results. Use the quick-drying formula on most washable surfaces, including chrome, stainless steel, bathroom fixtures and laminate countertops. It wipes clean and dries without smearing glass or mirror surfaces. Glass cleaner has no added fragrance and is super-concentrated for convenient use and improved efficiency. It comes in a J-Fill bottle that works with the J-Fill dispensing system for dispensing accurate cleaning solutions with no measuring, mixing or mess. Cleaner dispenses at a 1:64 dilution. Metering tip in every bottle ensures precise dilution, reduces maintenance and eliminates the possibility of tampering with dilution ratios. Simply push a button or squeeze the trigger.
Catalog No. 50-008-30675 Supplier Enterprise Technology Solutions Supplier No. DVO100975197
Super-concentrated, non-ammoniated glass cleaner meets EcoLogo and Greenguard standards and delivers streak-free results. Use the quick-drying formula on most washable surfaces, including chrome, stainless steel, bathroom fixtures and laminate countertops. It wipes clean and dries without smearing glass or mirror surfaces. Glass cleaner has no added fragrance and is super-concentrated for convenient use and improved efficiency. It comes in a J-Fill bottle that works with the J-Fill dispensing system for dispensing accurate cleaning solutions with no measuring, mixing or mess. Cleaner dispenses at a 1:64 dilution. Metering tip in every bottle ensures precise dilution, reduces maintenance and eliminates the possibility of tampering with dilution ratios. Simply push a button or squeeze the trigger.