Learn More
Enterprise Technology Solutions Diversey Care Cleaner,f/Glassware,Powder,Concentrated,Odorless,4lb,2/CT,RD
SDP
Catalog No. 5000831498
Encompass_Preferred
Beer Clean Glass Cleaner provides thoroughly cleaned glassware with no odor, aftertaste or residues for sparkling-clean glassware. Powdered compound cleans effectively in hot-cold, hard or soft water. Use with either manual or electric three-sink systems.
Catalog No. 50-008-31498 Supplier Enterprise Technology Solutions Supplier No. DVO95891201
Beer Clean Glass Cleaner provides thoroughly cleaned glassware with no odor, aftertaste or residues for sparkling-clean glassware. Powdered compound cleans effectively in hot-cold, hard or soft water. Use with either manual or electric three-sink systems.