Learn More
Enterprise Technology Solutions Diversey Care Cleaner,f/Toilets,Clinging,10%HCl,32oz,CitrusScnt,12/CT,GN
SDP
Catalog No. 5000790567
Encompass_Preferred
Crew Clinging Plus Toilet Bowl Cleaner effectively cleans surfaces and eliminates odors around toilet bowls and urinals. Specialty 10-percent HCl formula clings to toilet bowls and below the water line for tough stain removal. Cleaner leaves behind a refreshing citrus scent. Use on hard, inanimate, nonporous surfaces like floors, walls, porcelain and plastic to effectively clean in hospitals, nursing homes, schools, colleges and more.
Catalog No. 50-007-90567 Supplier Enterprise Technology Solutions Supplier No. DVO9302467
Crew Clinging Plus Toilet Bowl Cleaner effectively cleans surfaces and eliminates odors around toilet bowls and urinals. Specialty 10-percent HCl formula clings to toilet bowls and below the water line for tough stain removal. Cleaner leaves behind a refreshing citrus scent. Use on hard, inanimate, nonporous surfaces like floors, walls, porcelain and plastic to effectively clean in hospitals, nursing homes, schools, colleges and more.