Learn More
Enterprise Technology Solutions Diversey Care Cleaner, General Purpose, Hot Springs, 1 Gallon, 4/CT, YW
SDP
Catalog No. 5000831259
Encompass_Preferred
General-purpose cleaner features a unique formula that cleans without caustics, solvents or phosphates to easily clean unfinished tile in high-soil areas. Heavy-duty solution is great for spray-and-wipe, mop and bucket or auto-scrubber applications. Cleaner with a citrus scent is also perfect for scrubbing floors before re-coating finish.
Catalog No. 50-008-31259 Supplier Enterprise Technology Solutions Supplier No. DVO94039110
General-purpose cleaner features a unique formula that cleans without caustics, solvents or phosphates to easily clean unfinished tile in high-soil areas. Heavy-duty solution is great for spray-and-wipe, mop and bucket or auto-scrubber applications. Cleaner with a citrus scent is also perfect for scrubbing floors before re-coating finish.