Learn More
Enterprise Technology Solutions Diversey Care Cleaner, General Purpose, Hyd Per, 1.5 Gallon, 2/CT, Clear
SDP
Catalog No. 5000790905
Encompass_Preferred
Heavy-duty, concentrated cleaner features Accelerated Hydrogen Peroxide (AHP) technology for safe use on most hard surfaces, including marble floors. Multiple dilutions allow cleaning a wide range of cleaning applications, including floors, walls, glass and other hard surfaces. It can also be used for carpet extraction, pre-spray and spotting. Certified by EcoLogo and Green Seal cleaner does not contain any added dyes, phosphorus or fragrance. Colorless, odorless cleaner is perfect for auto scrubbers, mop and bucket and spray-and-wipe applications.
Catalog No. 50-007-90905 Supplier Enterprise Technology Solutions Supplier No. DVO94998832
Heavy-duty, concentrated cleaner features Accelerated Hydrogen Peroxide (AHP) technology for safe use on most hard surfaces, including marble floors. Multiple dilutions allow cleaning a wide range of cleaning applications, including floors, walls, glass and other hard surfaces. It can also be used for carpet extraction, pre-spray and spotting. Certified by EcoLogo and Green Seal cleaner does not contain any added dyes, phosphorus or fragrance. Colorless, odorless cleaner is perfect for auto scrubbers, mop and bucket and spray-and-wipe applications.