Learn More
Enterprise Technology Solutions Diversey Care Cleaner,Glass/MultiSurface,NoStreak,QuickDry,5L/RTD,Blue
SDP
Catalog No. 5000830992
Encompass_Preferred
Glance HC Glass and Multi-Surface Cleaner utilizes a non-streaking, quick-drying formula that effectively cleans and shines glass, mirrors, sinks and more. Easily cut through grease, grime, soap films, finger marks and smoke film. Use on glass and other washable surfaces like chrome, stainless steel, bathroom fixtures and laminate surfaces like countertops. Concentrated formula contains ammonia. Do not use on copper or brass, as repeated use may discolor these surfaces.
Catalog No. 50-008-30992 Supplier Enterprise Technology Solutions Supplier No. DVO93165337
Glance HC Glass and Multi-Surface Cleaner utilizes a non-streaking, quick-drying formula that effectively cleans and shines glass, mirrors, sinks and more. Easily cut through grease, grime, soap films, finger marks and smoke film. Use on glass and other washable surfaces like chrome, stainless steel, bathroom fixtures and laminate surfaces like countertops. Concentrated formula contains ammonia. Do not use on copper or brass, as repeated use may discolor these surfaces.