Learn More
Enterprise Technology Solutions Diversey Care Cleaner,J-Fill,No Rinse,Pine Scent,Kosher,5L,2/CT,Red
SDP
Catalog No. 5000831292
Encompass_Preferred
Spitfire SC Power Cleaner features a super-strength formula that eliminates grease, petroleum, tire marks and food soils. This powerful, solvent-based cleaner works on the toughest dirt, grime and stains on a wide variety of surfaces. Lightly scented with a fresh pine scent and pleasant to use, this no-rinse formula leaves no residue or streaks behind. Use this premium-performance surface cleaner for spray-and-wipe cleaning on hard surfaces, including removing ink, graffiti, heel marks and paint. This cleaner/degreaser is perfect for shops and kitchens. It comes in a J-Fill bottle that works with the J-Fill dispensing system for dispensing accurate cleaning solutions with no measuring, mixing or mess. Metering tip in every bottle ensures precise dilution, reduces maintenance and eliminates the possibility of tampering with dilution ratios. Simply push a button or squeeze the trigger.
Catalog No. 50-008-31292 Supplier Enterprise Technology Solutions Supplier No. DVO95892221
Spitfire SC Power Cleaner features a super-strength formula that eliminates grease, petroleum, tire marks and food soils. This powerful, solvent-based cleaner works on the toughest dirt, grime and stains on a wide variety of surfaces. Lightly scented with a fresh pine scent and pleasant to use, this no-rinse formula leaves no residue or streaks behind. Use this premium-performance surface cleaner for spray-and-wipe cleaning on hard surfaces, including removing ink, graffiti, heel marks and paint. This cleaner/degreaser is perfect for shops and kitchens. It comes in a J-Fill bottle that works with the J-Fill dispensing system for dispensing accurate cleaning solutions with no measuring, mixing or mess. Metering tip in every bottle ensures precise dilution, reduces maintenance and eliminates the possibility of tampering with dilution ratios. Simply push a button or squeeze the trigger.