Learn More
Enterprise Technology Solutions Diversey Care Cleaner,LowpH,RinseFree,Concentrated,Citrus,5Gal,Orange
SDP
Catalog No. 5000790719
Encompass_Preferred
Stride Citrus Neutral Cleaner is a non-alkaline formulation for everyday cleaning of floors and other hard surfaces. Concentrated, low-foaming formula allows less emptying of recovery tanks and features a low pH for a rinse-free clean that dries residue-free. Versatile cleaner performs in various application methods, like auto scrubber, spray-and-wipe, and mop-and-bucket. Enjoy a refreshing Citrus scent.
Catalog No. 50-007-90719 Supplier Enterprise Technology Solutions Supplier No. DVO101109754
Stride Citrus Neutral Cleaner is a non-alkaline formulation for everyday cleaning of floors and other hard surfaces. Concentrated, low-foaming formula allows less emptying of recovery tanks and features a low pH for a rinse-free clean that dries residue-free. Versatile cleaner performs in various application methods, like auto scrubber, spray-and-wipe, and mop-and-bucket. Enjoy a refreshing Citrus scent.