Learn More
Enterprise Technology Solutions Diversey Care Cleaner,Neutral,Concentrat,FragranceFree,2.5L/J-Fill,2/CT,OE
SDP
Catalog No. 5000831280
Encompass_Preferred
Stride Fragrance-Free Neutral Cleaner is a non-alkaline formulation for everyday cleaning of floors and other hard surfaces. Concentrated, low-foaming formula allows less emptying of recovery tanks and features a low pH for a rinse-free clean that dries residue-free. Versatile cleaner performs in various application methods, like auto scrubber, spray-and-wipe, and mop-and-bucket.
Catalog No. 50-008-31280 Supplier Enterprise Technology Solutions Supplier No. DVO94240626
Stride Fragrance-Free Neutral Cleaner is a non-alkaline formulation for everyday cleaning of floors and other hard surfaces. Concentrated, low-foaming formula allows less emptying of recovery tanks and features a low pH for a rinse-free clean that dries residue-free. Versatile cleaner performs in various application methods, like auto scrubber, spray-and-wipe, and mop-and-bucket.