Learn More
Enterprise Technology Solutions Diversey Care Cleaner,Neutral pH,Citrus,Multipurpose,5 Liter,Orange
SDP
Catalog No. 5000830459
Encompass_Preferred
Part of the Diversey RTD System, this neutral cleaner features a nonalkaline formula that dries film-free to prevent dulling floor finishes. Easy-to-use solution does not require rinsing for quick, simple cleaning performance. Low-foaming design allows less emptying of recovery tanks and increased performance. Concentrated cleaner with a citrus scent is Green Seal, Greenguard, NSF and Kosher certified. Neutral cleaner is perfect for everyday cleanings of nearly all washable surfaces in an autoscrubber, spray-and-wipe application or mop and bucket. Dilution ratio is 5:16.
Catalog No. 50-008-30459 Supplier Enterprise Technology Solutions Supplier No. DVO93063390
Part of the Diversey RTD System, this neutral cleaner features a nonalkaline formula that dries film-free to prevent dulling floor finishes. Easy-to-use solution does not require rinsing for quick, simple cleaning performance. Low-foaming design allows less emptying of recovery tanks and increased performance. Concentrated cleaner with a citrus scent is Green Seal, Greenguard, NSF and Kosher certified. Neutral cleaner is perfect for everyday cleanings of nearly all washable surfaces in an autoscrubber, spray-and-wipe application or mop and bucket. Dilution ratio is 5:16.