Learn More
Enterprise Technology Solutions Diversey Care Cleaner w/Hydrogen Peroxide, General-Purpose 1.4L, Clear
SDP
Catalog No. 5000831260
Encompass_Preferred
A concentrated, heavy-duty cleaner is safe to use on most hard floors and surfaces, including marble. It is based on proprietary Accelerated Hydrogen Peroxide (AHP) technology. Multiple dilutions allow cleaning a wide range of cleaning applications, including floors, walls, glass and other hard surfaces. PERdiem General-purpose Cleaner can be used for carpet extraction, pre-spray and spotting. Use in auto scrubbers, mop and bucket and spray-and-wipe applications. Bottle and pump are designed not to waste concentrate because they evacuate 99.9 percent of the liquid. Easy-to-use cleaner is colorless and odorless with no added dyes or fragrance. It is environmentally sustainable and recyclable. This PERdiem cleaner is part of the Diversey SmartDose system, effectively delivering accurate doses of concentrated product simply, safely and swiftly.
Catalog No. 50-008-31260 Supplier Enterprise Technology Solutions Supplier No. DVO95019481
A concentrated, heavy-duty cleaner is safe to use on most hard floors and surfaces, including marble. It is based on proprietary Accelerated Hydrogen Peroxide (AHP) technology. Multiple dilutions allow cleaning a wide range of cleaning applications, including floors, walls, glass and other hard surfaces. PERdiem General-purpose Cleaner can be used for carpet extraction, pre-spray and spotting. Use in auto scrubbers, mop and bucket and spray-and-wipe applications. Bottle and pump are designed not to waste concentrate because they evacuate 99.9 percent of the liquid. Easy-to-use cleaner is colorless and odorless with no added dyes or fragrance. It is environmentally sustainable and recyclable. This PERdiem cleaner is part of the Diversey SmartDose system, effectively delivering accurate doses of concentrated product simply, safely and swiftly.