Learn More
Enterprise Technology Solutions Diversey Care Cleanser,RTU,32 oz Squeeze Bottle,12/CT,Off-white
SDP
Catalog No. 5000790078
Encompass_Preferred
Mild acid formula quickly removes hard water deposits, grease, soap scum, scuff marks and other stubborn spots and buildup without scratching the surface. Emerel Multi-Surface Creme Cleanser clings to vertical surfaces so it stays where you put it to increase dwell time and therefore cleaning effectiveness. Ready-to-use cleanser in a squeeze bottle provides controlled sudsing so it rinses freely for a clean, bright surface. It is recommended for difficult to clean stains, including buildups, scuffs, grease, rubber marks and soap scum. Use on porcelain, ceramic, stainless steel, chrome, Formica, fiberglass and other washable surfaces not harmed by mild abrasives. Cleanser has a fresh scent.
Catalog No. 50-007-90078 Supplier Enterprise Technology Solutions Supplier No. DVO94995295
Mild acid formula quickly removes hard water deposits, grease, soap scum, scuff marks and other stubborn spots and buildup without scratching the surface. Emerel Multi-Surface Creme Cleanser clings to vertical surfaces so it stays where you put it to increase dwell time and therefore cleaning effectiveness. Ready-to-use cleanser in a squeeze bottle provides controlled sudsing so it rinses freely for a clean, bright surface. It is recommended for difficult to clean stains, including buildups, scuffs, grease, rubber marks and soap scum. Use on porcelain, ceramic, stainless steel, chrome, Formica, fiberglass and other washable surfaces not harmed by mild abrasives. Cleanser has a fresh scent.