Learn More
Enterprise Technology Solutions Diversey Care Degreaser/Cleaner,J-Fill,Conc,Solvent-free,2.5L,2/CT,Green
SDP
Catalog No. 5000790073
Encompass_Preferred
Tempest SC Solvent-free Cleaner/Degreaser removes food-based soils, oils and greases, metal working fluids and shop dirt. High-alkaline, solvent-free degreaser is made for industrial cleaning applications. It is suitable for use in mopping, autoscrubbing, steam cleaning, pressure-cleaning, foam cleaning vertical surfaces and immersion cleaning applications. Degreaser quickly penetrates industrial soils without solvents. Low-foaming, alkaline degreaser contains no VOCs. No fragrance is added. Cleaner/degreaser comes in a J-Fill bottle that works with the J-Fill dispensing system for dispensing accurate cleaning solutions with no measuring, mixing or mess. Metering tip in every bottle ensures precise dilution, reduces maintenance and eliminates the possibility of tampering with dilution ratios. Simply push a button or squeeze the trigger. Dilution ratios include 1:32 and 1:128.
Catalog No. 50-007-90073 Supplier Enterprise Technology Solutions Supplier No. DVO100986237
Tempest SC Solvent-free Cleaner/Degreaser removes food-based soils, oils and greases, metal working fluids and shop dirt. High-alkaline, solvent-free degreaser is made for industrial cleaning applications. It is suitable for use in mopping, autoscrubbing, steam cleaning, pressure-cleaning, foam cleaning vertical surfaces and immersion cleaning applications. Degreaser quickly penetrates industrial soils without solvents. Low-foaming, alkaline degreaser contains no VOCs. No fragrance is added. Cleaner/degreaser comes in a J-Fill bottle that works with the J-Fill dispensing system for dispensing accurate cleaning solutions with no measuring, mixing or mess. Metering tip in every bottle ensures precise dilution, reduces maintenance and eliminates the possibility of tampering with dilution ratios. Simply push a button or squeeze the trigger. Dilution ratios include 1:32 and 1:128.