Learn More
Enterprise Technology Solutions Diversey Care Degreaser,Concentrate,FoodSurface,HvyDty,2.5L/J-Fill,2/CT,YW
SDP
Catalog No. 5000790548
Encompass_Preferred
Suma Break-Up SC Heavy-Duty Degreaser quickly and easily cleans protein, fats, grease, oils and other food soils from surfaces without hard scrubbing. Highly alkaline, solvent-free formula is effective on many surfaces, including stainless steel, masonry, plastic, tile and all resilient and non-resilient floors (including sealed wood and cork). Concentrated degreaser is ideal for use in supermarket meat-cutting, seafood, bakery, deli and produce areas, institutional kitchens and food plants. Do not use on painted surfaces.
Catalog No. 50-007-90548 Supplier Enterprise Technology Solutions Supplier No. DVO95192347
Suma Break-Up SC Heavy-Duty Degreaser quickly and easily cleans protein, fats, grease, oils and other food soils from surfaces without hard scrubbing. Highly alkaline, solvent-free formula is effective on many surfaces, including stainless steel, masonry, plastic, tile and all resilient and non-resilient floors (including sealed wood and cork). Concentrated degreaser is ideal for use in supermarket meat-cutting, seafood, bakery, deli and produce areas, institutional kitchens and food plants. Do not use on painted surfaces.