Learn More
Enterprise Technology Solutions Diversey Care Dish Detergent,Liquid,f/Machines,5 Gal,Clear Red
SDP
Catalog No. 5000790578
Encompass_Preferred
Nonchlorinated, machine dish liquid is specially formulated to provide excellent soil removal in hard water conditions. It cuts through grease and dried-on food. Suma Nova L6 is suitable for use in a wide range of single and multi-tank, warewashing machines. It prevents scaling in dish-wash machines even in hard water condition. Dishwashing liquid should not be used with soft metals. Dilution ratios range from 1:400 to 1:1000.
Catalog No. 50-007-90578 Supplier Enterprise Technology Solutions Supplier No. DVO957252100
Nonchlorinated, machine dish liquid is specially formulated to provide excellent soil removal in hard water conditions. It cuts through grease and dried-on food. Suma Nova L6 is suitable for use in a wide range of single and multi-tank, warewashing machines. It prevents scaling in dish-wash machines even in hard water condition. Dishwashing liquid should not be used with soft metals. Dilution ratios range from 1:400 to 1:1000.