Learn More
Enterprise Technology Solutions Diversey Care DishwasherDetergent,HardWater,NonChlorinated,1Gal,4/CT,RD
SDP
Catalog No. 5000790077
Encompass_Preferred
Suma Nova L6 Dishwashing Detergent cuts through tough grease and dried-on food. Non-chlorinated dishwasher liquid is specially formulated to provide excellent soil removal while preventing scaling of your dishwasher, even in hard-water conditions. Suitable for use in a wide range of ware-washing machines, this dishwashing detergent is not for use with soft metals.
Catalog No. 50-007-90077 Supplier Enterprise Technology Solutions Supplier No. DVO957252280
Suma Nova L6 Dishwashing Detergent cuts through tough grease and dried-on food. Non-chlorinated dishwasher liquid is specially formulated to provide excellent soil removal while preventing scaling of your dishwasher, even in hard-water conditions. Suitable for use in a wide range of ware-washing machines, this dishwashing detergent is not for use with soft metals.