Learn More
Enterprise Technology Solutions Diversey Care Dispenser,Touchless/Manual,f/1.3L Soap/1.2L Sanitizer,WE
SDP
Catalog No. 5000831388
Encompass_Preferred
IntelliCare Hybrid Dispenser can address all your hand care needs with a versatile, modern and durable platform that is compatible with both liquid/gels and foam products. It dispenses 1300 ml. (1.3 liter) bottles of hand soap or 1200 ml. (1.2 liter) of alcohol-based hand sanitizer in a 0.4 ml. or 1 ml. dose per push. This hybrid dispenser also provides an automatic switch from touchless to manual if batteries die before replacement. When the refill is nearly empty, the locking dispenser will sound an alarm. It also has a low-battery signal. Translucent window allows good product visibility without product tampering. Dispenser is made of reliable, ABS plastic that is scratch-resistant and UV-resistant.
Catalog No. 50-008-31388 Supplier Enterprise Technology Solutions Supplier No. DVOD6205568
IntelliCare Hybrid Dispenser can address all your hand care needs with a versatile, modern and durable platform that is compatible with both liquid/gels and foam products. It dispenses 1300 ml. (1.3 liter) bottles of hand soap or 1200 ml. (1.2 liter) of alcohol-based hand sanitizer in a 0.4 ml. or 1 ml. dose per push. This hybrid dispenser also provides an automatic switch from touchless to manual if batteries die before replacement. When the refill is nearly empty, the locking dispenser will sound an alarm. It also has a low-battery signal. Translucent window allows good product visibility without product tampering. Dispenser is made of reliable, ABS plastic that is scratch-resistant and UV-resistant.