Learn More
Enterprise Technology Solutions Diversey Care Floor Cleaner,AccuMix,Concentrate,Citrus,32oz,Orange
SDP
Catalog No. 5000790530
Encompass_Preferred
Neutral cleaner concentrate is formulated for everyday cleaning of floors and other hard surfaces. It does not dull floor finishes. Nonalkaline cleaner dries film-free and does not require rinsing to deliver labor savings. Low-foaming formula with a citrus scent allows less emptying of recovery tanks. Use in spray-and-wipe applications, mop/bucket and autoscrubber. Dilution ratios include 1:128, 1:256, 1:384 and 1:768. Floor cleaner is part of the AccuMix cleaning system so it comes with a built-in accurate measuring spout to get the right proportions every time.
Catalog No. 50-007-90530 Supplier Enterprise Technology Solutions Supplier No. DVO101109755
Neutral cleaner concentrate is formulated for everyday cleaning of floors and other hard surfaces. It does not dull floor finishes. Nonalkaline cleaner dries film-free and does not require rinsing to deliver labor savings. Low-foaming formula with a citrus scent allows less emptying of recovery tanks. Use in spray-and-wipe applications, mop/bucket and autoscrubber. Dilution ratios include 1:128, 1:256, 1:384 and 1:768. Floor cleaner is part of the AccuMix cleaning system so it comes with a built-in accurate measuring spout to get the right proportions every time.