Learn More
Enterprise Technology Solutions Diversey Care Floor Cleaner,Deodorizer,Concentrated,High-Gloss,Damage-Free,No-Rinse,Citrus Scent,100/BG,1/CT,OE
SDP
Catalog No. 5000790486
Encompass_Preferred
Aqua Chempacs Citrus Floor Cleaner is a concentrated, deodorizing floor cleaner specially formulated for the daily maintenance of high-gloss floors. It quickly removes dirt, grease and grime without dulling, hazing or marring the original finish. It also effectively cleans walls, counters, equipment and painted areas. For ultimate convenience, this cleaner requires no rinsing and leaves surfaces film-free. It deodorizes surfaces and leaves behind a fresh, citrus scent.
Catalog No. 50-007-90486 Supplier Enterprise Technology Solutions Supplier No. DVO9320930
Aqua Chempacs Citrus Floor Cleaner is a concentrated, deodorizing floor cleaner specially formulated for the daily maintenance of high-gloss floors. It quickly removes dirt, grease and grime without dulling, hazing or marring the original finish. It also effectively cleans walls, counters, equipment and painted areas. For ultimate convenience, this cleaner requires no rinsing and leaves surfaces film-free. It deodorizes surfaces and leaves behind a fresh, citrus scent.