Learn More
Enterprise Technology Solutions Diversey Care Floor Cleaner,Heavy Duty,Citrus,32 oz,YW
SDP
Catalog No. 5000789905
Encompass_Preferred
Prominence Heavy Duty Floor Cleaner is a highly effective cleaner designed to remove soils and oils without dulling or altering the appearance of your floor. Super-concentrated formula delivers a powerful cleaning performance. It quickly emulsifies dirt to remove soils and dries film-free with no rinsing required. Formula is pH neutral when diluted. Cleaner is Greenguard certified and meets the Green Seal GS-37 standard for industrial and institutional use. It works as a daily floor cleaner on most floor types or as a high-performance, general-purpose cleaner.
Catalog No. 50-007-89905 Supplier Enterprise Technology Solutions Supplier No. DVO94996440
Prominence Heavy Duty Floor Cleaner is a highly effective cleaner designed to remove soils and oils without dulling or altering the appearance of your floor. Super-concentrated formula delivers a powerful cleaning performance. It quickly emulsifies dirt to remove soils and dries film-free with no rinsing required. Formula is pH neutral when diluted. Cleaner is Greenguard certified and meets the Green Seal GS-37 standard for industrial and institutional use. It works as a daily floor cleaner on most floor types or as a high-performance, general-purpose cleaner.