Learn More
Enterprise Technology Solutions Diversey Care Floor Cleaner,HeavyDuty,f/DailyMaintenance,Concentrated,LavenderScent,100/PK,1/CT,Purple
SDP
Catalog No. 5000790425
Encompass_Preferred
This Aqua Chempacs Heavy-Duty Floor Cleaner thoroughly cleans dirt and traffic soils, including black heel marks. Intended for daily maintenance, this concentrated formula is mild enough for daily cleaning, yet effective enough for deep scrubbing at stronger dilutions. It leaves behind a refreshing lavender scent with each clean.
Catalog No. 50-007-90425 Supplier Enterprise Technology Solutions Supplier No. DVO9320928
This Aqua Chempacs Heavy-Duty Floor Cleaner thoroughly cleans dirt and traffic soils, including black heel marks. Intended for daily maintenance, this concentrated formula is mild enough for daily cleaning, yet effective enough for deep scrubbing at stronger dilutions. It leaves behind a refreshing lavender scent with each clean.