Learn More
Enterprise Technology Solutions Diversey Care Floor Cleaner,Hvy-Dty,Pro,No-Rinse,Citrus,2.5L,2/CT,YW
Supplier: Enterprise Technology Solutions DVO94996466
Highly effective cleaner is designed to remove soils and oils without dulling or altering the appearance of your floor. Prominence Heavy-duty Floor Cleaner quickly emulsifies dirt for fast removal and is designed for use on most floor types. It is pH neutral when diluted. This daily floor cleaner offers the flexibility of multiple dilution ratios (1:256; 1:513) to handle all types of soil loads. It is part of a comprehensive floor care offering for cleaning, maintaining, stripping and finishing floors. Floors dry film-free, and no rinsing is required. Greenguard approved, Green Seal certified cleaner is clear yellow-green in color and has a citrus scent. It comes in a J-Fill bottle that works with the J-Fill dispensing system for dispensing accurate cleaning solutions with no measuring, mixing or mess. Metering tip in every bottle ensures precise dilution, reduces maintenance and eliminates the possibility of tampering with dilution ratios. Simply push a button or squeeze the trigger.