Learn More
Enterprise Technology Solutions Diversey Care Floor Cleaner, Hvy-Dty, Pro, No-Rinse, Citrus, 5L, Yellow
SDP
Catalog No. 5000790080
Encompass_Preferred
Part of this Diversey RTD System, this highly effective cleaner is designed to remove soils and oils without dulling or altering the appearance of your floor. Prominence Heavy-duty Floor Cleaner quickly emulsifies dirt for fast removal and is designed for use on most floor types. It is pH neutral when diluted. This daily floor cleaner offers the flexibility of multiple dilution ratios (1:256; 1:513) to handle all types of soil loads. It is part of a comprehensive floor care offering for cleaning, maintaining, stripping and finishing floors. Floors dry film-free, and no rinsing is required. Greenguard approved, Green Seal certified cleaner is clear yellow-green in color and has a citrus scent.
Catalog No. 50-007-90080 Supplier Enterprise Technology Solutions Supplier No. DVO94996458
Part of this Diversey RTD System, this highly effective cleaner is designed to remove soils and oils without dulling or altering the appearance of your floor. Prominence Heavy-duty Floor Cleaner quickly emulsifies dirt for fast removal and is designed for use on most floor types. It is pH neutral when diluted. This daily floor cleaner offers the flexibility of multiple dilution ratios (1:256; 1:513) to handle all types of soil loads. It is part of a comprehensive floor care offering for cleaning, maintaining, stripping and finishing floors. Floors dry film-free, and no rinsing is required. Greenguard approved, Green Seal certified cleaner is clear yellow-green in color and has a citrus scent.