Learn More
Enterprise Technology Solutions Diversey Care Floor Cleaner, Neutral, Raindance, 1.5 Gallon, 2/CT, Yellow
SDP
Catalog No. 5000790895
Encompass_Preferred
Neutral floor cleaner features a powerful, concentrated formula to effectively clean floors and other hard surfaces. The low-foam formula is perfect for use in auto scrubbers and in mop-and-bucket applications. Nonalkaline solution will not dull high-gloss finishes. Easy-to-use design does not require rinsing for fast, cost-effective cleaning. Floor cleaner with a floral scent is Green Seal and Greenguard certified.
Catalog No. 50-007-90895 Supplier Enterprise Technology Solutions Supplier No. DVO93323981
Neutral floor cleaner features a powerful, concentrated formula to effectively clean floors and other hard surfaces. The low-foam formula is perfect for use in auto scrubbers and in mop-and-bucket applications. Nonalkaline solution will not dull high-gloss finishes. Easy-to-use design does not require rinsing for fast, cost-effective cleaning. Floor cleaner with a floral scent is Green Seal and Greenguard certified.